Learn More
Biomatik Corporation Tumor Necrosis Factor Receptor Superfamily, Member 21 (TNFRSF21) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU22988
Encompass_Preferred
Tumor Necrosis Factor Receptor Superfamily, Member 21 (Tnfrsf21) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Tumor Necrosis Factor Receptor Superfamily, Member 21 (TNFRSF21). The Tumor Necrosis Factor Receptor Superfamily, Member 21 (Tnfrsf21) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Lys406~His605. Applications: WB, IHC, ICC, IP. Target Synonyms: Tumor Necrosis Factor Receptor Superfamily, Member 21 (TNFRSF21). Accession Number: Q9EPU5. Gene ID: 94185. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9803