Learn More
Biomatik Corporation Lipolysis Stimulated Lipoprotein Receptor (LSR) anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU23127
Encompass_Preferred
Lipolysis Stimulated Lipoprotein Receptor (Lsr) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Lipolysis Stimulated Lipoprotein Receptor (LSR). The Lipolysis Stimulated Lipoprotein Receptor (Lsr) Polyclonal Antibody has been validated for the following applications: WB, ICC/IF. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Ser35~Glu205. Applications: WB, ICC/IF. Target Synonyms: Lipolysis Stimulated Lipoprotein Receptor (LSR). Accession Number: Q99KG5. Gene ID: 54135. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9805