Learn More
Biomatik Corporation Cyclin A1 (CCNA1) anti-Human, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509808
Encompass_Preferred
The Cyclin A1 (CCNA1) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: CCNA1 (Ser183-Tyr430). Applications: WB, IHC. Alternative Names: CCN-A1. UniProt: P78396. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9808 Supplier Biomatik Corporation Supplier No. CAU23451
The Cyclin A1 (CCNA1) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: CCNA1 (Ser183-Tyr430). Applications: WB, IHC. Alternative Names: CCN-A1. UniProt: P78396. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.