Learn More
Biomatik Corporation Collagen Type III Alpha 1 (COL3a1) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509811
Encompass_Preferred
The Collagen Type III Alpha 1 (COL3a1) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse. Immunogen: COL3a1 (Gly434-Pro643). Applications: WB, IHC. Alternative Names: COL3-A1; COL3A1; EDS4A; Collagen Type III Alpha 1; Ehlers-Danlos Syndrome Type IV Autosomal Dominant; Collagen Alpha-1 (III)chain. UniProt: Q7TT32. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9811 Supplier Biomatik Corporation Supplier No. CAU23492
The Collagen Type III Alpha 1 (COL3a1) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse. Immunogen: COL3a1 (Gly434-Pro643). Applications: WB, IHC. Alternative Names: COL3-A1; COL3A1; EDS4A; Collagen Type III Alpha 1; Ehlers-Danlos Syndrome Type IV Autosomal Dominant; Collagen Alpha-1 (III)chain. UniProt: Q7TT32. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.