Learn More
Biomatik Corporation Carbonic Anhydrase IX (CA9) anti-Human, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU23602
Encompass_Preferred
The Carbonic Anhydrase IX (CA9) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: CA9 (Pro59-Asp414). Applications: WB, IHC. Alternative Names: CA-IX; CAIX; MN; pMW1; P54/58N; Carbonic Dehydratase; Renal cell carcinoma-associated antigen G250; RCC-Associated Protein G250; Carbonate dehydratase IX. UniProt: Q16790. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9814