Learn More
Biomatik Corporation Adhesion Regulating Molecule 1 (ADRM1) anti-Human, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU24459
Encompass_Preferred
The Adhesion Regulating Molecule 1 (ADRM1) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: ADRM1 (Thr2-Asp407). Applications: WB, IHC. Alternative Names: GP110; Rpn13; Proteasomal Ubiquitin Receptor ADRM1; 110 kDa cell membrane glycoprotein; Proteasome regulatory particle non-ATPase 13. UniProt: Q16186. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9834