Learn More
Biomatik Corporation Hepatocyte Growth Factor Receptor (HGFR) anti-Human, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509835
Encompass_Preferred
The Hepatocyte Growth Factor Receptor (HGFR) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: HGFR (Val1092-Val1379). Applications: WB, IHC. Alternative Names: CMET; RCCP2; C-Met; Met Proto-Oncogene; Mesenchymal-Epithelial Transition Factor; HGF/SF receptor; Proto-oncogene c-Met; Scatter factor receptor; Tyrosine-protein kinase Met. UniProt: P08581. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9835 Supplier Biomatik Corporation Supplier No. CAU24571
The Hepatocyte Growth Factor Receptor (HGFR) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: HGFR (Val1092-Val1379). Applications: WB, IHC. Alternative Names: CMET; RCCP2; C-Met; Met Proto-Oncogene; Mesenchymal-Epithelial Transition Factor; HGF/SF receptor; Proto-oncogene c-Met; Scatter factor receptor; Tyrosine-protein kinase Met. UniProt: P08581. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.