Learn More
Biomatik Corporation Collagen Type VIII Alpha 1 (COL8a1) anti-Human, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU24615
Encompass_Preferred
The Collagen Type VIII Alpha 1 (COL8a1) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: COL8a1 (Pro575-Pro743). Applications: WB, IHC. Alternative Names: COL8-A1; Collagen Alpha-1 (VIII)chain; Endothelial collagen; Vastatin. UniProt: P27658. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9838