Learn More
Biomatik Corporation VEGF Co Regulated Chemokine 1 (VCC1) anti-Human, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509844
Encompass_Preferred
Vegf Co Regulated Chemokine 1 (Vcc1) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to VEGF Co Regulated Chemokine 1 (VCC1). The Vegf Co Regulated Chemokine 1 (Vcc1) Polyclonal Antibody has been validated for the following applications: WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Leu24~Leu119. Applications: WB. Target Synonyms: VEGF Co Regulated Chemokine 1 (VCC1). Accession Number: Q6UXB2. Gene ID: 284340. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9844 Supplier Biomatik Corporation Supplier No. CAU24753
Vegf Co Regulated Chemokine 1 (Vcc1) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to VEGF Co Regulated Chemokine 1 (VCC1). The Vegf Co Regulated Chemokine 1 (Vcc1) Polyclonal Antibody has been validated for the following applications: WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Leu24~Leu119. Applications: WB. Target Synonyms: VEGF Co Regulated Chemokine 1 (VCC1). Accession Number: Q6UXB2. Gene ID: 284340. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.