Learn More
Biomatik Corporation Ki-67 Protein (Ki67P) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU24782
Encompass_Preferred
Ki-67 Protein (Ki-67) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Ki-67 Protein (Ki-67). The Ki-67 Protein (Ki-67) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Met1~Cys247. Applications: WB, IHC, ICC, IP. Target Synonyms: Ki-67 Protein (Ki-67). Accession Number: E9PVX6. Gene ID: 17345. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9846