Learn More
Biomatik Corporation Apolipoprotein A5 (APOA5) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509850
Encompass_Preferred
The Apolipoprotein A5 (APOA5) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse. Immunogen: APOA5 (Arg21-Pro210). Applications: WB, IHC. Alternative Names: Apo-A5; APOA-V; APOAV; RAP3; Regeneration-associated protein 3. UniProt: Q8C7G5. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9850 Supplier Biomatik Corporation Supplier No. CAU24880
The Apolipoprotein A5 (APOA5) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse. Immunogen: APOA5 (Arg21-Pro210). Applications: WB, IHC. Alternative Names: Apo-A5; APOA-V; APOAV; RAP3; Regeneration-associated protein 3. UniProt: Q8C7G5. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.