Learn More
Biomatik Corporation Hemojuvelin (HJV) anti-Human, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509851
Encompass_Preferred
The Hemojuvelin (HJV) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: HJV (Lys234-Ser416). Applications: WB, IHC. Alternative Names: HFE2; HFE2A; HJV; JH; RGMC; hemochromatosis type 2 (Juvenile); Haemojuvelin; RGM domain family member C. UniProt: Q6ZVN8. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9851 Supplier Biomatik Corporation Supplier No. CAU24888
The Hemojuvelin (HJV) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: HJV (Lys234-Ser416). Applications: WB, IHC. Alternative Names: HFE2; HFE2A; HJV; JH; RGMC; hemochromatosis type 2 (Juvenile); Haemojuvelin; RGM domain family member C. UniProt: Q6ZVN8. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.