Learn More
Biomatik Corporation Complement Component 8g (C8g) anti-Human, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU24961
Encompass_Preferred
The Complement Component 8g (C8g) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: C8g (Lys39-Arg202). Applications: WB, IHC. UniProt: P07360. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9853