Learn More
Biomatik Corporation Leucine Rich Alpha-2-Glycoprotein 1 (LRG1) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509854
Encompass_Preferred
Leucine Rich Alpha-2-Glycoprotein 1 (Lrg1) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Leucine Rich Alpha-2-Glycoprotein 1 (LRG1). The Leucine Rich Alpha-2-Glycoprotein 1 (Lrg1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Ser136~Leu342. Applications: WB, IHC, ICC, IP. Target Synonyms: Leucine Rich Alpha-2-Glycoprotein 1 (LRG1). Accession Number: Q91XL1. Gene ID: 76905. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9854 Supplier Biomatik Corporation Supplier No. CAU25040
Leucine Rich Alpha-2-Glycoprotein 1 (Lrg1) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Leucine Rich Alpha-2-Glycoprotein 1 (LRG1). The Leucine Rich Alpha-2-Glycoprotein 1 (Lrg1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Ser136~Leu342. Applications: WB, IHC, ICC, IP. Target Synonyms: Leucine Rich Alpha-2-Glycoprotein 1 (LRG1). Accession Number: Q91XL1. Gene ID: 76905. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.