Learn More
Biomatik Corporation Glyceraldehyde-3-Phosphate Dehydrogenase (GAPDH) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU25045
Encompass_Preferred
The Glyceraldehyde-3-Phosphate Dehydrogenase (GAPDH) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse. Immunogen: GAPDH (Gly56-Phe284). Applications: WB, IHC. Alternative Names: G3PD; GAPD; Peptidyl-cysteine S-nitrosylase GAPDH. UniProt: P16858. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9855