Learn More
Biomatik Corporation Interleukin 7 Receptor (IL7R) anti-Human, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU25091
Encompass_Preferred
Interleukin 7 Receptor (Il7R) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Interleukin 7 Receptor (IL7R). The Interleukin 7 Receptor (Il7R) Polyclonal Antibody has been validated for the following applications: WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Lys173~Ala260. Applications: WB, IHC. Target Synonyms: Interleukin 7 Receptor (IL7R). Accession Number: P16871. Gene ID: 3575. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9857