Learn More
Biomatik Corporation Forkhead Box Protein P3 (FOXP3) anti-Human, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU25137
Encompass_Preferred
The Forkhead Box Protein P3 (FOXP3) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: FOXP3 (Tyr191-Glu412). Applications: WB, IHC. Alternative Names: FOX-P3; AIID; DIETER; IPEX; JM2; PIDX; XPID; SCURFIN; Immune Dysregulation,Polyendocrinopathy,Enteropathy,X-Linked. UniProt: Q9BZS1. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9861