Learn More
Biomatik Corporation Nuclear Factor Kappa B (NFkB) anti-Human, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU25233
Encompass_Preferred
Nuclear Factor Kappa B (Nfkb) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Nuclear Factor Kappa B (NFkB). The Nuclear Factor Kappa B (Nfkb) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Pro42~Met367. Applications: WB, IHC, ICC, IP. Target Synonyms: Nuclear Factor Kappa B (NFkB). Accession Number: P19838. Gene ID: 4790. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9869