Learn More
Biomatik Corporation Serum Amyloid A2 (SAA2) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU25292
Encompass_Preferred
Serum Amyloid A2 (Saa2) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Serum Amyloid A2 (SAA2). The Serum Amyloid A2 (Saa2) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Gly20~Tyr122. Applications: WB, IHC, ICC, IP. Target Synonyms: Serum Amyloid A2 (SAA2). Accession Number: P05367. Gene ID: 20209. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9871