Learn More
Biomatik Corporation Cluster of Differentiation (CD163) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU25430
Encompass_Preferred
The Cluster of Differentiation (CD163) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse. Immunogen: CD163 (Pro697-Gly972). Applications: WB, IHC. Alternative Names: M130; MM130; Scavenger receptor cysteine-rich type 1 protein M130; Hemoglobin scavenger receptor. UniProt: Q2VLH6. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9875