Learn More
Biomatik Corporation Defensin Alpha 1, Neutrophil (DEFa1) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU25461
Encompass_Preferred
The Defensin Alpha 1, Neutrophil (DEFa1) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse. Immunogen: DEFa1 (Asp20-Arg93). Applications: WB, IHC. Alternative Names: DEF1; DEFA2; HNP-1; HP-1; MRS; DEF3; HNP-3; HNP3; HP-3; Defensin,Alpha 1; Myeloid-Related Sequence. UniProt: P11477. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9876