Learn More
Biomatik Corporation Glycophorin A (GYPA) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU25463
Encompass_Preferred
The Glycophorin A (GYPA) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse. Immunogen: GYPA (Met1-Val108). Applications: WB, IHC. Alternative Names: CD235a; MN; MNS; GPA; GPErik; GPSAT; PAS-2; GpMiIII; HGpMiIII; HGpMiV; HGpMiX; HGpMiXI; HGpSta; Sialoglycoprotein alpha; MN sialoglycoprotein. UniProt: P14220. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9877