Learn More
Biomatik Corporation Cholesterol (CH) anti-General, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU25467
Encompass_Preferred
The Cholesterol (CH) Polyclonal Antibody is raised in Rabbit. Species Reactivity: General. Immunogen: Small Molecule, Cholesterol conjugated to OVA. Applications: WB, IHC, ICC, IP. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9878