Learn More
Biomatik Corporation Myostatin (MSTN) anti-Human, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509881
Encompass_Preferred
Myostatin (Mstn) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Myostatin (MSTN). The Myostatin (Mstn) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Asp267~Ser375. Applications: WB, IHC, ICC, IP. Target Synonyms: Myostatin (MSTN). Accession Number: O14793. Gene ID: 2660. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9881 Supplier Biomatik Corporation Supplier No. CAU25555
Myostatin (Mstn) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Myostatin (MSTN). The Myostatin (Mstn) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Asp267~Ser375. Applications: WB, IHC, ICC, IP. Target Synonyms: Myostatin (MSTN). Accession Number: O14793. Gene ID: 2660. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.