Learn More
Biomatik Corporation Prominin 1 (PROM1) anti-Human, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509887
Encompass_Preferred
Prominin 1 (Prom1) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Prominin 1 (PROM1). The Prominin 1 (Prom1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Ile171~Asp431. Applications: WB, IHC, ICC, IP. Target Synonyms: Prominin 1 (PROM1). Accession Number: O43490. Gene ID: 8842. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9887 Supplier Biomatik Corporation Supplier No. CAU25781
Prominin 1 (Prom1) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Prominin 1 (PROM1). The Prominin 1 (Prom1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Ile171~Asp431. Applications: WB, IHC, ICC, IP. Target Synonyms: Prominin 1 (PROM1). Accession Number: O43490. Gene ID: 8842. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.