Learn More
Biomatik Corporation Hemoglobin (HB) anti-Bovine, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU25987
Encompass_Preferred
The Hemoglobin (HB) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Bovine. Immunogen: Native Protein Hemoglobin. Applications: WB. Alternative Names: Hgb; Haemoglobin; Heterotetramer (αβ)2. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9892