Learn More
Biomatik Corporation Vascular Endothelial Growth Factor Receptor 2 (VEGFR2) anti-Human, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU26065
Encompass_Preferred
Vascular Endothelial Growth Factor Receptor 2 (Vegfr2) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Vascular Endothelial Growth Factor Receptor 2 (VEGFR2). The Vascular Endothelial Growth Factor Receptor 2 (Vegfr2) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Asn46~Thr320. Applications: WB, IHC, ICC, IP. Target Synonyms: Vascular Endothelial Growth Factor Receptor 2 (VEGFR2). Accession Number: P35968. Gene ID: 3791. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9893