Learn More
Biomatik Corporation Complement Component 4b (C4b) anti-Human, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509897
Encompass_Preferred
The Complement Component 4b (C4b) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: C4b (Asn680-Arg756). Applications: WB, IHC. Alternative Names: CPAMD3; C4F; CO4; C4B1; C4B3; CH; C4 (Chido Blood Group); Basic complement C4; C4a anaphylatoxin; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 3. UniProt: P0C0L5. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9897 Supplier Biomatik Corporation Supplier No. CAU26178
The Complement Component 4b (C4b) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: C4b (Asn680-Arg756). Applications: WB, IHC. Alternative Names: CPAMD3; C4F; CO4; C4B1; C4B3; CH; C4 (Chido Blood Group); Basic complement C4; C4a anaphylatoxin; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 3. UniProt: P0C0L5. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.