Learn More
Biomatik Corporation Cytotoxic T-Lymphocyte Associated Antigen 4 (CTLA4) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU26313
Encompass_Preferred
The Cytotoxic T-Lymphocyte Associated Antigen 4 (CTLA4) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse. Immunogen: CTLA4 (Val45-Ser185). Applications: WB, IHC. Alternative Names: CD152; CELIAC3; CTLA4; GSE; IDDM12; Celiac Disease 3; Cytotoxic T-lymphocyte protein 4. UniProt: P09793. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9904