Learn More
Biomatik Corporation Integrin Alpha X (ITGaX) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU26439
Encompass_Preferred
Integrin Alpha X (Cd11C) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Integrin Alpha X (CD11c). The Integrin Alpha X (Cd11C) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Arg907~Val1115. Applications: WB, IHC, ICC, IP. Target Synonyms: Integrin Alpha X (CD11c). Accession Number: Q9QXH4. Gene ID: 16411. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9911