Learn More
Biomatik Corporation Deoxyribonuclease I (DNASE1) anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509912
Encompass_Preferred
The Deoxyribonuclease I (DNASE1) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: DNASE1 (Gly19-Ala259). Applications: WB, IHC. Alternative Names: DNaseI; DNL1; DRNI; DNase-I; Dornase alfa. UniProt: P24855. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9912 Supplier Biomatik Corporation Supplier No. CAU26474
The Deoxyribonuclease I (DNASE1) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: DNASE1 (Gly19-Ala259). Applications: WB, IHC. Alternative Names: DNaseI; DNL1; DRNI; DNase-I; Dornase alfa. UniProt: P24855. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.