Learn More
Biomatik Corporation Interleukin 12 Receptor Beta 2 (IL12Rb2) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509917
Encompass_Preferred
Interleukin 12 Receptor Beta 2 (Il12Rb2) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Interleukin 12 Receptor Beta 2 (IL12Rb2). The Interleukin 12 Receptor Beta 2 (Il12Rb2) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Met285~Gln441. Applications: WB, IHC, ICC, IP. Target Synonyms: Interleukin 12 Receptor Beta 2 (IL12Rb2). Accession Number: P97378. Gene ID: 16162. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9917 Supplier Biomatik Corporation Supplier No. CAU26563
Interleukin 12 Receptor Beta 2 (Il12Rb2) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Interleukin 12 Receptor Beta 2 (IL12Rb2). The Interleukin 12 Receptor Beta 2 (Il12Rb2) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Met285~Gln441. Applications: WB, IHC, ICC, IP. Target Synonyms: Interleukin 12 Receptor Beta 2 (IL12Rb2). Accession Number: P97378. Gene ID: 16162. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.