Learn More
Biomatik Corporation Integrin Beta 1 (ITGb1) anti-Human, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509921
Encompass_Preferred
Integrin Beta 1 (Itgb1) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Integrin Beta 1 (ITGb1). The Integrin Beta 1 (Itgb1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Asp140~Ile383. Applications: WB, IHC, ICC, IP. Target Synonyms: Integrin Beta 1 (ITGb1). Accession Number: P05556. Gene ID: 3688. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9921 Supplier Biomatik Corporation Supplier No. CAU26617
Integrin Beta 1 (Itgb1) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Integrin Beta 1 (ITGb1). The Integrin Beta 1 (Itgb1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Asp140~Ile383. Applications: WB, IHC, ICC, IP. Target Synonyms: Integrin Beta 1 (ITGb1). Accession Number: P05556. Gene ID: 3688. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.