Learn More
Biomatik Corporation Vimentin (VIM) anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU26621
Encompass_Preferred
Vimentin (Vim) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Vimentin (VIM). The Vimentin (Vim) Polyclonal Antibody has been validated for the following applications: WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Ser2~Glu466. Applications: WB, IHC, IF. Target Synonyms: Vimentin (VIM). Accession Number: P08670. Gene ID: 7431. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9922