Learn More
Biomatik Corporation Glypican 1 (GPC1) anti-Human, Pig, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU26634
Encompass_Preferred
The Glypican 1 (GPC1) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Pig. Immunogen: GPC1 (Leu114-Lys277). Applications: WB. Alternative Names: Glypican Proteoglycan 1; Secreted glypican-1. UniProt: P35052. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9924