Learn More
Biomatik Corporation Melatonin (MT) anti-General, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509935
Encompass_Preferred
Melatonin (Mt) Polyclonal Antibody is a Universal Polyclonal Antibody to Melatonin (MT). The Melatonin (Mt) Polyclonal Antibody has been validated for the following applications: ELISA, CLIA / IHC-Fr, ICC, IP (predicted). Host: Rabbit. Species: Universal. Applications: ELISA, CLIA / IHC-Fr, ICC, IP (predicted). Target Synonyms: Melatonin (MT). Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9935 Supplier Biomatik Corporation Supplier No. CAU26842
Melatonin (Mt) Polyclonal Antibody is a Universal Polyclonal Antibody to Melatonin (MT). The Melatonin (Mt) Polyclonal Antibody has been validated for the following applications: ELISA, CLIA / IHC-Fr, ICC, IP (predicted). Host: Rabbit. Species: Universal. Applications: ELISA, CLIA / IHC-Fr, ICC, IP (predicted). Target Synonyms: Melatonin (MT). Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.