Learn More
Biomatik Corporation Gonadotropin Releasing Hormone (GnRH) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU26974
Encompass_Preferred
The Gonadotropin Releasing Hormone (GnRH) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse. Immunogen: GnRH (Gln22-Met90). Applications: WB, IHC. Alternative Names: GNRH1; GRH; LNRH; LHRH; LH-RH I; Luliberin; Luteinizing-Hormone Releasing Hormone; Progonadoliberin-1; Gonadoliberin I; Gonadorelin; GnRH-associated peptide 1. UniProt: P12025. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9942