Learn More
Biomatik Corporation Von Willebrand Factor (vWF) anti-Pig, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU27008
Encompass_Preferred
Von Willebrand Factor (Vwf) Polyclonal Antibody is a Pig (Sus scrofa; Porcine) Polyclonal Antibody to Von Willebrand Factor (vWF). The Von Willebrand Factor (Vwf) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Pig (Sus scrofa; Porcine). Immunogen: Asp1167~Val1334. Applications: WB, IHC, ICC, IP. Target Synonyms: Von Willebrand Factor (vWF). Accession Number: Q28833. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9943