Learn More
Biomatik Corporation Von Willebrand Factor (vWF) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509944
Encompass_Preferred
Von Willebrand Factor (Vwf) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Von Willebrand Factor (vWF). The Von Willebrand Factor (Vwf) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Asp1498~Val1665. Applications: WB, IHC, ICC, IP. Target Synonyms: Von Willebrand Factor (vWF). Accession Number: Q8CIZ8. Gene ID: 22371. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9944 Supplier Biomatik Corporation Supplier No. CAU27009
Von Willebrand Factor (Vwf) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Von Willebrand Factor (vWF). The Von Willebrand Factor (Vwf) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Asp1498~Val1665. Applications: WB, IHC, ICC, IP. Target Synonyms: Von Willebrand Factor (vWF). Accession Number: Q8CIZ8. Gene ID: 22371. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.