Learn More
Biomatik Corporation 5-Hydroxytryptamine (5-HT) anti-General, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU27083
Encompass_Preferred
The 5-Hydroxytryptamine (5-HT) Polyclonal Antibody is raised in Rabbit. Species Reactivity: General. Immunogen: Small Molecule, 5-HT conjugated to OVA. Applications: WB, IHC, ICC, IP. Alternative Names: 5HT; Serotonin; N-Methyl-Gamma. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9947