Learn More
Biomatik Corporation Hypoxia Inducible Factor 1 Alpha (HIF1a) anti-Human, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509949
Encompass_Preferred
Hypoxia Inducible Factor 1 Alpha (Hif1A) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Hypoxia Inducible Factor 1 Alpha (HIF1a). The Hypoxia Inducible Factor 1 Alpha (Hif1A) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Thr218~Thr506. Applications: WB, IHC, ICC, IP. Target Synonyms: Hypoxia Inducible Factor 1 Alpha (HIF1a). Accession Number: Q16665. Gene ID: 3091. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9949 Supplier Biomatik Corporation Supplier No. CAU27106
Hypoxia Inducible Factor 1 Alpha (Hif1A) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Hypoxia Inducible Factor 1 Alpha (HIF1a). The Hypoxia Inducible Factor 1 Alpha (Hif1A) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Thr218~Thr506. Applications: WB, IHC, ICC, IP. Target Synonyms: Hypoxia Inducible Factor 1 Alpha (HIF1a). Accession Number: Q16665. Gene ID: 3091. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.