Learn More
Biomatik Corporation Programmed Cell Death Protein 1 Ligand 2 (PDCD1LG2) anti-Human, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU27135
Encompass_Preferred
Programmed Cell Death Protein 1 Ligand 2 (Pdl2) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Programmed Cell Death Protein 1 Ligand 2 (PDL2). The Programmed Cell Death Protein 1 Ligand 2 (Pdl2) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Glu33~Phe236. Applications: WB, IHC, ICC, IP. Target Synonyms: Programmed Cell Death Protein 1 Ligand 2 (PDL2). Accession Number: Q9BQ51. Gene ID: 80380. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9952