Learn More
Biomatik Corporation Programmed Cell Death Protein 1 Ligand 1 (PDCD1LG1) anti-Mouse, Rat, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU27138
Encompass_Preferred
Programmed Cell Death Protein 1 Ligand 1 (Pdl1) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Programmed Cell Death Protein 1 Ligand 1 (PDL1). The Programmed Cell Death Protein 1 Ligand 1 (Pdl1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Leu48~Thr290. Applications: WB, IHC, ICC, IP. Target Synonyms: Programmed Cell Death Protein 1 Ligand 1 (PDL1). Accession Number: Q9EP73. Gene ID: 60533. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9954