Learn More
Biomatik Corporation Lactoferrin (LTF) anti-Human, Dog, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU27173
Encompass_Preferred
Lactoferrin (Ltf) Polyclonal Antibody is a Dog (Canis familiaris; Canine) Polyclonal Antibody to Lactoferrin (LTF). The Lactoferrin (Ltf) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Dog (Canis familiaris; Canine). Immunogen: Cys28~Lys351. Applications: WB, IHC, ICC, IP. Target Synonyms: Lactoferrin (LTF). Accession Number: A0A8I3NMR3. Gene ID: 484787. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9957