Learn More
Biomatik Corporation Programmed Cell Death Protein 1 (PDCD1) anti-Pig, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU27234
Encompass_Preferred
Programmed Cell Death Protein 1 (Pd1) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Programmed Cell Death Protein 1 (PD1). The Programmed Cell Death Protein 1 (Pd1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC/IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Leu41~Ala132. Applications: WB, IHC, ICC/IF. Target Synonyms: Programmed Cell Death Protein 1 (PD1). Accession Number: Q15116. Gene ID: 5133. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9962