Learn More
Biomatik Corporation Complement Receptor 2 (CR2) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509963
Encompass_Preferred
The Complement Receptor 2 (CR2) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse. Immunogen: CR2 (Cys463-Asp665). Applications: WB, IHC. Alternative Names: CD21; C3DR; Complement Component (3d/Epstein Barr Virus)Receptor 2; Complement C3d receptor; Epstein-Barr virus receptor; EBV receptor. UniProt: P19070. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9963 Supplier Biomatik Corporation Supplier No. CAU27236
The Complement Receptor 2 (CR2) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse. Immunogen: CR2 (Cys463-Asp665). Applications: WB, IHC. Alternative Names: CD21; C3DR; Complement Component (3d/Epstein Barr Virus)Receptor 2; Complement C3d receptor; Epstein-Barr virus receptor; EBV receptor. UniProt: P19070. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.