Learn More
Biomatik Corporation Complement Factor H (CFH) anti-Rat, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU27476
Encompass_Preferred
The Complement Factor H (CFH) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Rat. Immunogen: CFH (Ala975-Ser1217). Applications: WB, IHC. Alternative Names: CF-H; FH; FHL1; ARMD4; ARMS1; CFHL3; HF1; HF2; HUS; H Factor 2; Age-Related Maculopathy Susceptibility 1; Adrenomedullin binding protein. UniProt: G3V9R2. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9971