Learn More
Biomatik Corporation Caspase 9 (CASP9) anti-Human, Rat, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU27499
Encompass_Preferred
The Caspase 9 (CASP9) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Rat. Immunogen: CASP9 (Ala331-Ser416). Applications: WB, IHC. Alternative Names: APAF3; ICE-LAP6; MCH6; Apoptosis-Related Cysteine Peptidase; Cysteinyl Aspartate Specific Proteinases 9; Apoptotic protease-activating factor 3; ICE-like apoptotic protease 6. UniProt: P55211. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9974