Learn More
Biomatik Corporation Caspase 3 (CASP3) anti-Human, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509977
Encompass_Preferred
The Caspase 3 (CASP3) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: CASP3 (Ser29-Asp175). Applications: WB, IHC. Alternative Names: CPP32; CPP32B; SCA1; Apoptain; Yama; Apoptosis-Related Cysteine Peptidase; Cysteinyl Aspartate Specific Proteinases 3; SREBP cleavage activity 1. UniProt: P42574. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9977 Supplier Biomatik Corporation Supplier No. CAU27505
The Caspase 3 (CASP3) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: CASP3 (Ser29-Asp175). Applications: WB, IHC. Alternative Names: CPP32; CPP32B; SCA1; Apoptain; Yama; Apoptosis-Related Cysteine Peptidase; Cysteinyl Aspartate Specific Proteinases 3; SREBP cleavage activity 1. UniProt: P42574. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.