Learn More
Biomatik Corporation V-Rel Reticuloendotheliosis Viral Oncogene Homolog A (RELA) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509980
Encompass_Preferred
Transcription Factor P65 (Nfkb3) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Transcription Factor P65 (RELA). The Transcription Factor P65 (Nfkb3) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Pro19~Tyr306. Applications: WB, IHC, ICC, IP. Target Synonyms: Transcription Factor P65 (RELA). Accession Number: Q04207. Gene ID: 19697. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9980 Supplier Biomatik Corporation Supplier No. CAU27518
Transcription Factor P65 (Nfkb3) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Transcription Factor P65 (RELA). The Transcription Factor P65 (Nfkb3) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Pro19~Tyr306. Applications: WB, IHC, ICC, IP. Target Synonyms: Transcription Factor P65 (RELA). Accession Number: Q04207. Gene ID: 19697. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.